Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259374.2 | NP_001246303.1 | 5 - 6 | 1394 | 66 | |
NM_001259375.2 | NP_001246304.1 | 5 - 6 | 1785 | 66 | ||
NM_137058.4 | NP_610902.1 | 5 - 6 | 1785 | 66 | ||
GenBank mRNA | AY069830.1 | - | 5 - 6 | 2105 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |