Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stj | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259374.2 | NP_001246303.1 | 7 - 8 | 1982 | 87 | |
NM_001259375.2 | NP_001246304.1 | 7 - 8 | 2373 | 87 | ||
NM_137058.4 | NP_610902.1 | 7 - 8 | 2373 | 87 | ||
GenBank mRNA | AY069830.1 | - | 7 - 8 | 2693 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |