Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DJ-1alpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137072.1 | NP_610916.1 | 2 - 3 | 516 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |