Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299473.1 | NP_001286402.1 | 6 - 7 | 2327 | 89 | |
NM_137081.5 | NP_610925.3 | 6 - 7 | 2327 | 89 | ||
GenBank mRNA | BT044467.1 | - | 6 - 7 | 1410 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |