Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259386.2 | NP_001246315.2 | 7 - 8 | 2392 | 82 | |
NM_137081.5 | NP_610925.3 | 8 - 9 | 2587 | 82 | ||
GenBank mRNA | BT044467.1 | - | 8 - 9 | 1670 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |