Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ir52d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137198.2 | NP_611042.2 | 1 | 1736 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |