Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp4E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169198.2 | NP_001162669.1 | 8 - 9 | 4828 | 67 | |
NM_001169199.2 | NP_001162670.1 | 8 - 9 | 4746 | 67 | ||
NM_001169200.2 | NP_001162671.2 | 8 - 9 | 4724 | 67 | ||
NM_001169201.2 | NP_001162672.1 | 8 - 9 | 4747 | 67 | ||
NM_001297946.1 | NP_001284875.1 | 8 - 9 | 4724 | 67 | ||
NM_080337.3 | NP_525076.2 | 8 - 9 | 4724 | 67 | ||
NM_167024.3 | NP_726967.1 | 8 - 9 | 4724 | 67 | ||
GenBank mRNA | BT033064.1 | - | 8 - 9 | 4719 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |