Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042796.3 | NP_001036261.3 | 20 - 21 | 6852 | 61 | |
NM_001144685.2 | NP_001138157.2 | 20 - 21 | 7164 | 61 | ||
NM_001144686.2 | NP_001138158.2 | 20 - 21 | 7194 | 61 | ||
NM_001144687.2 | NP_001138159.2 | 19 - 20 | 6573 | 61 | ||
NM_001297949.1 | NP_001284878.1 | 23 - 24 | 11181 | 61 | ||
NM_001297950.1 | NP_001284879.1 | 20 - 21 | 7194 | 61 | ||
NM_167028.3 | NP_726978.2 | 20 - 21 | 7904 | 61 |
Gene Symbol: CG15465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297948.1 | NP_001284877.1 | 23 - 24 | 11181 | 61 | |
NM_131978.3 | NP_572206.1 | 20 - 21 | 7904 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |