Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hex-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272455.2 | NP_001259384.1 | 1 | 2467 | 132 | |
NM_080109.4 | NP_524848.1 | 1 | 2252 | 132 | ||
NM_167194.3 | NP_727350.1 | 1 | 2252 | 132 | ||
GenBank mRNA | AY069391.1 | - | 1 | 2257 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |