Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: X11Lbeta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272496.2 | NP_001259425.2 | 5 - 6 | 4490 | 77 | |
NM_167239.4 | NP_727440.3 | 7 - 8 | 3572 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |