Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tango5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169245.2 | NP_001162716.1 | 4 - 5 | 1259 | 66 | |
NM_001272501.1 | NP_001259430.1 | 4 - 5 | 2193 | 66 | ||
NM_001298157.1 | NP_001285086.1 | 4 - 5 | 1350 | 66 | ||
NM_167242.3 | NP_727444.1 | 4 - 5 | 1174 | 66 | ||
NM_167243.3 | NP_727445.1 | 3 - 4 | 1367 | 66 | ||
NM_167244.3 | NP_727446.1 | 4 - 5 | 942 | 66 | ||
GenBank mRNA | BT133079.1 | - | 4 - 5 | 1157 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |