Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab9Fb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167261.3 | NP_727472.1 | 1 | 628 | 92 | |
GenBank mRNA | BT024998.1 | - | 1 | 628 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |