Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klp10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298180.1 | NP_001285109.1 | 3 - 4 | 1717 | 79 | |
NM_132459.4 | NP_572687.1 | 3 - 4 | 1629 | 79 | ||
NM_167267.2 | NP_727491.1 | 3 - 4 | 2004 | 79 | ||
NM_167268.2 | NP_727492.1 | 2 - 3 | 1793 | 79 | ||
NM_167269.2 | NP_727493.1 | 3 - 4 | 1625 | 79 | ||
NM_167270.2 | NP_727494.1 | 3 - 4 | 1629 | 79 | ||
GenBank mRNA | AY051763.1 | - | 3 - 4 | 1622 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |