Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132466.3 | NP_572694.1 | 2 - 3 | 443 | 59 | |
NM_167274.2 | NP_727501.1 | 2 - 3 | 443 | 59 | ||
GenBank mRNA | AY058302.1 | - | 2 - 3 | 426 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |