Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp4g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132493.3 | NP_572721.1 | 5 - 6 | 2093 | 70 | |
NM_167287.2 | NP_727531.2 | 5 - 6 | 2093 | 70 | ||
GenBank mRNA | AY060719.1 | - | 5 - 6 | 2070 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |