Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Karl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298208.1 | NP_001285137.1 | 2 - 3 | 1287 | 62 | |
NM_001298209.1 | NP_001285138.1 | 1 - 2 | 614 | 62 | ||
NM_132520.4 | NP_572748.2 | 1 - 2 | 258 | 62 | ||
NM_167303.3 | NP_727559.1 | 1 - 2 | 258 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |