Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258496.3 | NP_001245425.1 | 8 - 9 | 5434 | 80 | |
NM_001272130.1 | NP_001259059.1 | 8 - 9 | 5437 | 80 | ||
NM_079884.5 | NP_524623.2 | 8 - 9 | 5437 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |