Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mGluR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272147.1 | NP_001259076.1 | 6 - 7 | 1924 | 70 | |
NM_001272148.1 | NP_001259077.1 | 5 - 6 | 2262 | 70 | ||
NM_001297789.1 | NP_001284718.1 | 5 - 6 | 1380 | 70 | ||
NM_079900.3 | NP_524639.2 | 6 - 7 | 1924 | 70 | ||
GenBank mRNA | BT088899.1 | - | 5 - 6 | 2236 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |