Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF4G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103382.2 | NP_001096852.1 | 9 - 10 | 4513 | 80 | |
NM_001144385.2 | NP_001137857.1 | 8 - 9 | 3984 | 80 | ||
NM_079901.4 | NP_524640.3 | 8 - 9 | 3754 | 80 | ||
GenBank mRNA | AF030155.1 | - | 8 - 9 | 3760 | 80 | |
BT053671.1 | - | 8 - 9 | 3748 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |