Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258662.2 | NP_001245591.1 | 1 | 904 | 154 | |
NM_079905.3 | NP_524644.1 | 1 | 1115 | 154 | ||
GenBank mRNA | AF181623.1 | - | 1 | 1115 | 154 | |
AF217797.2 | - | 1 | 883 | 154 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |