Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298883.1 | NP_001285812.1 | 5 - 6 | 1323 | 63 | |
NM_079928.2 | NP_524667.1 | 5 - 6 | 836 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |