Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299510.1 | NP_001286439.1 | 4 - 5 | 1427 | 58 | |
NM_079963.4 | NP_524702.3 | 4 - 5 | 1427 | 58 | ||
NM_176175.2 | NP_788355.1 | 4 - 5 | 1427 | 58 | ||
GenBank mRNA | AY121698.1 | - | 4 - 5 | 1425 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |