Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PpD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079968.3 | NP_524707.1 | 1 | 1213 | 129 | |
GenBank mRNA | BT133328.1 | - | 1 | 1150 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster |