Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ScpX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079976.6 | NP_524715.2 | 4 - 5 | 1558 | 81 | |
GenBank mRNA | AY089539.1 | - | 4 - 5 | 1561 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |