Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sut3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273869.1 | NP_001260798.1 | 2 - 3 | 901 | 71 | |
NM_079992.4 | NP_524731.1 | 2 - 3 | 909 | 71 | ||
GenBank mRNA | AF199485.1 | - | 2 - 3 | 881 | 71 | |
BT044406.1 | - | 2 - 3 | 909 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |