Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Roc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297825.1 | NP_001284754.1 | 1 - 2 | 226 | 61 | |
NM_130496.4 | NP_569852.1 | 1 - 2 | 226 | 61 | ||
GenBank mRNA | AY119265.1 | - | 1 - 2 | 243 | 61 | |
KX531597.1 | - | 1 - 2 | 82 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |