Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp52F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137249.4 | NP_611093.2 | 11 - 12 | 2415 | 67 | |
GenBank mRNA | BT133387.1 | - | 11 - 12 | 2395 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |