Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptp52F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137249.4 | NP_611093.2 | 14 - 15 | 3561 | 66 | |
GenBank mRNA | BT133387.1 | - | 14 - 15 | 3541 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |