Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274092.1 | NP_001261021.1 | 3 - 4 | 3621 | 83 | |
NM_137251.3 | NP_611095.1 | 3 - 4 | 3908 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |