Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eIF3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299616.1 | NP_001286545.1 | 2 - 3 | 266 | 64 | |
NM_137398.2 | NP_611242.1 | 2 - 3 | 185 | 64 | ||
GenBank mRNA | BT044242.1 | - | 2 - 3 | 266 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster Inventoried |