Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: swi2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137403.4 | NP_611247.3 | 2 - 3 | 557 | 65 | |
GenBank mRNA | AY084149.1 | - | 2 - 3 | 555 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |