Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jheh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137542.4 | NP_611386.2 | 3 - 4 | 698 | 72 | |
NM_176233.4 | NP_788413.4 | 3 - 4 | 1003 | 72 | ||
GenBank mRNA | AB107959.1 | - | 3 - 4 | 672 | 72 | |
AF517546.1 | - | 3 - 4 | 608 | 72 | ||
BT044122.1 | - | 3 - 4 | 698 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |