Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jheh3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137543.4 | NP_611387.1 | 1 - 2 | 557 | 68 | |
GenBank mRNA | AY061427.1 | - | 1 - 2 | 529 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |