Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259488.2 | NP_001246417.1 | 3 - 4 | 2222 | 57 | |
NM_001274157.1 | NP_001261086.1 | 3 - 4 | 2166 | 57 | ||
NM_137564.3 | NP_611408.2 | 3 - 4 | 2166 | 57 | ||
GenBank mRNA | BT010287.1 | - | 3 - 4 | 2125 | 57 | |
BT100309.1 | - | 3 - 4 | 2306 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |