Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mRpL54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137697.4 | NP_611541.1 | 1 - 2 | 231 | 90 | |
GenBank mRNA | BT044303.1 | - | 1 - 2 | 196 | 90 | |
KX531867.1 | - | 1 - 2 | 107 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |