Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272572.2 | NP_001259501.1 | 9 - 10 | 4369 | 62 | |
NM_001382110.1 | NP_001368981.1 | 8 - 9 | 3573 | 62 | ||
NM_001382111.1 | NP_001368982.1 | 8 - 9 | 3573 | 62 | ||
NM_167346.2 | NP_727661.1 | 8 - 9 | 3573 | 62 | ||
GenBank mRNA | AY069695.1 | - | 8 - 9 | 3508 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |