Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IP3K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132647.2 | NP_572875.1 | 2 - 3 | 1136 | 132 | |
NM_167358.2 | NP_727688.1 | 2 - 3 | 1260 | 132 | ||
GenBank mRNA | BT133057.1 | - | 2 - 3 | 1134 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |