Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mud | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258736.2 | NP_001245665.1 | 2 - 3 | 606 | 85 | |
NM_001258737.2 | NP_001245666.1 | 2 - 3 | 606 | 85 | ||
NM_001272615.2 | NP_001259544.1 | 2 - 3 | 606 | 85 | ||
NM_001272616.1 | NP_001259545.1 | 2 - 3 | 606 | 85 | ||
NM_080495.6 | NP_536743.3 | 2 - 3 | 606 | 85 | ||
NM_167399.6 | NP_727769.3 | 2 - 3 | 606 | 85 | ||
NM_167400.5 | NP_727770.2 | 2 - 3 | 606 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |