Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272624.2 | NP_001259553.1 | 3 - 4 | 375 | 90 | |
NM_001272626.2 | NP_001259555.1 | 3 - 4 | 716 | 90 | ||
NM_167415.3 | NP_727797.1 | 3 - 4 | 375 | 90 | ||
NM_167416.3 | NP_727798.1 | 3 - 4 | 487 | 90 | ||
GenBank mRNA | BT126133.1 | - | 3 - 4 | 354 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |