Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: drd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038756.2 | NP_001033845.1 | 8 - 9 | 2534 | 75 | |
GenBank mRNA | BT021240.1 | - | 8 - 9 | 2532 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |