Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sep4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_132919.3 | NP_573147.2 | 2 - 3 | 840 | 76 | |
NM_167530.2 | NP_728003.1 | 2 - 3 | 388 | 76 | ||
NM_167531.2 | NP_728004.1 | 2 - 3 | 526 | 76 | ||
NM_167532.2 | NP_728005.1 | 2 - 3 | 863 | 76 | ||
NM_167533.2 | NP_728006.1 | 2 - 3 | 859 | 76 | ||
NM_167534.2 | NP_728007.1 | 2 - 3 | 694 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |