Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vav | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103560.3 | NP_001097030.1 | 6 - 7 | 1933 | 85 | |
NM_001272774.1 | NP_001259703.1 | 5 - 6 | 1822 | 85 | ||
NM_133144.4 | NP_573372.1 | 5 - 6 | 1309 | 85 | ||
NM_167642.3 | NP_728235.1 | 5 - 6 | 1309 | 85 | ||
GenBank mRNA | AY069536.1 | - | 5 - 6 | 1298 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |