Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kek5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258817.2 | NP_001245746.1 | 4 - 5 | 2681 | 83 | |
NM_133154.5 | NP_573382.1 | 5 - 6 | 2644 | 83 | ||
NM_167647.3 | NP_728242.1 | 5 - 6 | 2714 | 83 | ||
NM_167648.3 | NP_728243.1 | 7 - 8 | 2814 | 83 | ||
GenBank mRNA | BT023770.1 | - | 5 - 6 | 2698 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |