Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CoRest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014754.1 | NP_001014754.1 | 7 - 8 | 1138 | 88 | |
NM_001014755.2 | NP_001014755.1 | 6 - 7 | 1423 | 88 | ||
NM_001014756.2 | NP_001014756.1 | 6 - 7 | 1177 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |