Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CoRest | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014752.4 | NP_001014752.2 | 8 - 9 | 2072 | 97 | |
NM_001014753.3 | NP_001014753.1 | 8 - 9 | 2066 | 97 | ||
NM_001201742.3 | NP_001188671.1 | 10 - 11 | 2774 | 97 | ||
NM_001258824.1 | NP_001245753.1 | 10 - 11 | 2854 | 97 | ||
NM_001258825.2 | NP_001245754.1 | 10 - 11 | 2954 | 97 | ||
NM_001258826.1 | NP_001245755.1 | 3 - 4 | 979 | 97 | ||
GenBank mRNA | BT023817.1 | - | 8 - 9 | 2050 | 97 | |
BT099711.1 | - | 10 - 11 | 2678 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Drosophila melanogaster |