Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mri | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299954.1 | NP_001286883.1 | 3 - 4 | 1400 | 77 | |
NM_138164.4 | NP_612008.1 | 4 - 5 | 1311 | 77 | ||
NM_167801.3 | NP_728484.1 | 3 - 4 | 1403 | 77 | ||
GenBank mRNA | BT099621.1 | - | 4 - 5 | 1254 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |