Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mthl14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_167821.3 | NP_728509.1 | 6 - 7 | 1381 | 108 | |
GenBank mRNA | AY071287.1 | - | 6 - 7 | 1377 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |