Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fwd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274274.1 | NP_001261203.1 | 12 - 13 | 5790 | 101 | |
NM_001370043.1 | NP_001356971.1 | 11 - 12 | 6023 | 101 | ||
NM_080083.4 | NP_524822.1 | 11 - 12 | 6023 | 101 | ||
NM_167826.2 | NP_728518.1 | 11 - 12 | 5267 | 101 | ||
NM_167827.2 | NP_728519.1 | 10 - 11 | 4096 | 101 | ||
GenBank mRNA | AF242375.2 | - | 10 - 11 | 4096 | 101 | |
AY052049.1 | - | 11 - 12 | 6004 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |