Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mur2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201595.2 | NP_001188524.1 | 4 - 5 | 1162 | 72 | |
NM_001297860.1 | NP_001284789.1 | 5 - 6 | 1523 | 72 | ||
NM_130571.3 | NP_569927.1 | 4 - 5 | 1162 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |