Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PsGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169175.2 | NP_001162646.1 | 2 - 3 | 1136 | 60 | |
NM_001169176.2 | NP_001162647.1 | 2 - 3 | 1136 | 60 | ||
NM_001272258.1 | NP_001259187.1 | 2 - 3 | 1136 | 60 | ||
NM_001272259.1 | NP_001259188.1 | 2 - 3 | 1136 | 60 | ||
NM_001272260.1 | NP_001259189.1 | 2 - 3 | 1136 | 60 | ||
NM_130645.6 | NP_570001.5 | 2 - 3 | 1136 | 60 | ||
NM_130646.3 | NP_570002.1 | 2 - 3 | 1136 | 60 | ||
NM_206610.5 | NP_996333.4 | 2 - 3 | 1136 | 60 | ||
GenBank mRNA | BT024192.1 | - | 2 - 3 | 1137 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |